cis-1,3-Cyclopentanedicarboxylic acid 95%

Supplier: Ambeed
Warning

Synonyms: cis-Cyclopentane-1,3-dicarboxylic acid

A186933-250MG A186933-1G A186933-5G
103815-980EA 28 USD
103815-980 103846-568 76985-954 77084-258 77084-260
cis-1,3-Cyclopentanedicarboxylic acid 95%
cis-1,3-Cyclopentanedicarboxylic acid

Precaución: for Research Use Only

Formula: C₇H₁₀O₄
MW: 158.16 g/mol
Boiling Pt: 320.7 °C (760 mm Hg)
Flash Pt: 161.9 °C
Storage Temperature: Ambient
MDL Number: MFCD01311175
CAS Number: 876-05-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR