cis-1,4-Cyclohexanedicarboxylic acid 90%

Supplier: Ambeed
Warning

Synonyms: cis-Cyclohexane-1,4-dicarboxylic acid

A400182-100G A400182-10G A400182-250MG A400182-1G A400182-25G A400182-5G
103827-042EA 7 USD
103827-042 103838-076 77195-926 77195-928 77195-930 77307-066 77307-068 77307-070
cis-1,4-Cyclohexanedicarboxylic acid 90%
cis-1,4-Cyclohexanedicarboxylic acid

Precaución: for Research Use Only

Formula: C₈H₁₂O₄
MW: 172.1800 g/mol
Boiling Pt: 384.1 °C (760 mm Hg)
Melting Pt: 167…176 °C
Flash Pt: 200.254 °C
Storage Temperature: Ambient
MDL Number: MFCD00064950
CAS Number: 619-81-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR