cis-1,3-Cyclohexanedicarboxylic acid 95%

Supplier: Ambeed
Warning

Synonyms: cis-Cyclohexane-1,3-dicarboxylic acid

A107542-250MG A107542-1G A107542-25G
103797-208EA 20 USD
103797-208 103822-976 77160-554 77283-630 77682-986
cis-1,3-Cyclohexanedicarboxylic acid 95%
cis-1,3-Cyclohexanedicarboxylic acid

Precaución: For research use only.

Formula: C₈H₁₂O₄
MW: 172.1800 g/mol
Boiling Pt: 332.4 °C (760 mm Hg)
Melting Pt: 165 °C
Flash Pt: 169 °C
Storage Temperature: Ambient
MDL Number: MFCD00134411
CAS Number: 2305-31-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR