Cis-4-Hydroxy-D-proline hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: (2R,4R)-4-Hydroxypyrrolidine-2-carboxylic acid hydrochloride, H-cis-D-Hyp-OH·HCl

A116148-10G A116148-25G A116148-5G A116148-100G
103824-546EA 8 USD
103824-546 77136-276 77136-278 77311-366 77614-914
Cis-4-Hydroxy-D-proline hydrochloride 95%
Cis-4-Hydroxy-D-proline hydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₀ClNO₃
MW: 167.5908 g/mol
Boiling Pt: 368.6 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD09749867
CAS Number: 77449-94-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR