cis-4-Aminocyclohexanol hydrochloride 97%

Supplier: Ambeed
Warning

A353503-100G A353503-10G A353503-1G A353503-25G A353503-5G
103826-096EA 17 USD
103826-096 103837-098 76817-346 76817-348 77306-642 77306-644 77306-646
cis-4-Aminocyclohexanol hydrochloride 97%
cis-4-Aminocyclohexanol hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₄ClNO
Boiling Pt: 241.2 °C (760 mm Hg)
Melting Pt: 225…227 °C
Flash Pt: 75.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07366531
CAS Number: 56239-26-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR