cis-4-Aminocyclohexanecarboxylic acid 97%

Supplier: Ambeed
Warning

A628291-1G A628291-100G A628291-10G A628291-250MG A628291-25G A628291-5G
103840-264EA 54 USD
103840-264 76844-428 76980-662 76980-664 76980-666 77308-002 77308-004
cis-4-Aminocyclohexanecarboxylic acid 97%
cis-4-Aminocyclohexanecarboxylic acid

Precaución: For research use only

Formula: C₇H₁₃NO₂
Melting Pt: 299…301 °C
Flash Pt: 123.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00191730
CAS Number: 3685-23-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR