cis-3-Aminocyclohexanecarboxylic acid 90%

Supplier: Ambeed
Warning

A179732-10G A179732-1G A179732-25G
103807-458EA 13 USD
103807-458 103845-810 76974-404 77301-626 77301-628
cis-3-Aminocyclohexanecarboxylic acid 90%
cis-3-Aminocyclohexanecarboxylic acid

Precaución: for Research Use Only

Formula: C₇H₁₃NO₂
MW: 143.19 g/mol
Boiling Pt: 280 °C (760 mm Hg)
Flash Pt: 123.1 °C
Storage Temperature: Ambient
MDL Number: MFCD09753647
CAS Number: 16636-51-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR