cis-2-Phenyl-1,3-dioxan-5-ol 97%

Supplier: Ambeed
Warning

A158197-250MG A158197-1G A158197-5G
103805-104EA 18 USD
103805-104 103843-312 76689-684 77319-164 77319-166
cis-2-Phenyl-1,3-dioxan-5-ol 97%
cis-2-Phenyl-1,3-dioxan-5-ol

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₃
MW: 180.2000 g/mol
Boiling Pt: 333.6 °C (760 mm Hg)
Melting Pt: 83…86 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00274343
CAS Number: 4141-19-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR