Racemic cis-2-Fluorocyclopropanecarboxylic acid 97%

Supplier: Ambeed
Danger

Synonyms: cis-2-Fluorocyclopropanecarboxylic acid

A104724-10G A104724-250MG A104724-1G
103796-766EA 24 USD
103796-766 103822-490 76809-512 76809-514 77283-264
Racemic cis-2-Fluorocyclopropanecarboxylic acid 97%
Racemic cis-2-Fluorocyclopropanecarboxylic acid

Precaución: for Research Use Only

Formula: C₄H₅FO₂
MW: 104.08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04972869
CAS Number: 105919-34-4
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR