cis-2-Aminocyclohexanol hydrochloride 95%

Supplier: Ambeed
Warning

A229214-100MG A229214-250MG
103819-454EA 315 USD
103819-454 76692-038 77879-389
cis-2-Aminocyclohexanol hydrochloride 95%
cis-2-Aminocyclohexanol hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₄ClNO
Storage Temperature: Ambient
MDL Number: MFCD00143981
CAS Number: 6936-47-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR