Cilastatin 95%

Supplier: Ambeed
Warning

Synonyms: (Z)-7-(((R)-2-Amino-2-carboxyethyl)thio)-2-((S)-2,2-dimethylcyclopropane-1-carboxamido)hept-2-enoic acid

A371776-10G A371776-250MG A371776-1G A371776-5G
103792-680EA 11 USD
103792-680 103799-630 103826-492 103837-510 76694-342 76694-344 77306-842 77306-844
Cilastatin 95%
Cilastatin

Precaución: for Research Use Only

Formula: C₁₆H₂₆N₂O₅S
MW: 358.45 g/mol
Boiling Pt: 655.5 °C (760 mm Hg)
Flash Pt: 350.2 °C
Storage Temperature: Freezer
MDL Number: MFCD00867379
CAS Number: 82009-34-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR