CID-16020046 98%

Supplier: Ambeed
Warning

Synonyms: 4-(4-(3-Hydroxyphenyl)-6-oxo-3-(p-tolyl)-4,6-dihydropyrrolo[3,4-c]pyrazol-5(1H)-yl)benzoic acid

A843612-1MG A843612-10MG A843612-50MG A843612-5MG
103791-732EA 80 USD
103791-732 103792-860 103792-862 103799-728 77244-476 77308-440 77308-442 77308-444
CID-16020046 98%
CID-16020046

Precaución: for Research Use Only

Formula: C₂₅H₁₉N₃O₄
MW: 425.43 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06196075
CAS Number: 834903-43-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR