4-Acetamido-N-(2-aminophenyl)benzamide 98%

Supplier: Ambeed
Warning

A132717-1MG A132717-10MG A132717-100MG A132717-250MG
103792-128EA 19 USD
103792-128 103799-342 103801-772 76996-480 77298-574 77599-906 77670-882
4-Acetamido-N-(2-aminophenyl)benzamide 98%
4-Acetamido-N-(2-aminophenyl)benzamide

Precaución: for Research Use Only

Formula: C₁₅H₁₅N₃O₂
Flash Pt: 226.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00866266
CAS Number: 112522-64-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR