Chrysin 98%

Supplier: Ambeed
Warning

Synonyms: 5,7-Dihydroxy-2-phenyl-4H-chromen-4-one, 5,7-Dihydroxyflavone, 5,7-Dihydroxy-2-phenyl-4H-benzo[b]pyran-4-one, 5,7-Dihydroxy-2-phenyl-4H-1-benzopyran-4-one, 5,7-Dihydroxy-2-phenyl-4-benzopyrone, 5,7-Dihydroxy-2-phenylchromen-4-one

A771467-100G A771467-100MG A771467-10G A771467-500G A771467-25G A771467-5G A771467-1G
103840-936EA 11 USD
103840-936 76791-480 76791-482 76791-484 76791-486 76845-184 76845-186 77308-294
Chrysin 98%
Chrysin

Precaución: for Research Use Only

Formula: C₁₅H₁₀O₄
MW: 254.24 g/mol
Boiling Pt: 429
Melting Pt: 284…286 °C
Flash Pt: 192.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00006834
CAS Number: 480-40-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR