5,7-Dichloro-2-methyl-8-quinolinol 98%

Supplier: Ambeed
Warning

Synonyms: 5,7-Dichloro-8-hydroxy-2-methylquinoline, 5,7-Dichloro-2-methylquinolin-8-ol

A140767-100G A140767-100MG A140767-10G A140767-1G A140767-25G A140767-5G
76811-754EA 212.18 USD
76811-754 76811-756 76811-758 76811-760 77299-160 77316-014
5,7-Dichloro-2-methyl-8-quinolinol 98%
5,7-Dichloro-2-methyl-8-quinolinol

Precaución: for Research Use Only

Formula: C₁₀H₇Cl₂NO
MW: 228.0747 g/mol
Boiling Pt: 350.7 °C (760 mm Hg)
Melting Pt: 108…114 °C
Flash Pt: 165.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00023984
CAS Number: 72-80-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR