Chlorotris(triphenylphosphine)cobalt(I) 98%

Supplier: Ambeed
Warning

A241454-250MG A241454-1G A241454-25G A241454-5G
103811-410EA 8 USD
103811-410 103851-214 76692-294 77090-380 77304-212 77304-214
Chlorotris(triphenylphosphine)cobalt(I) 98%
Chlorotris(triphenylphosphine)cobalt(I)

Precaución: for Research Use Only

Formula: C₅₄H₄₅ClCoP₃
MW: 881.26 g/mol
Melting Pt: 135…139 °C
Storage Temperature: Ambient
MDL Number: MFCD00015864
CAS Number: 26305-75-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR