Chloromethyl benzoate 98%

Supplier: Ambeed
Warning

A176570-10G A176570-1G A176570-5G
103807-150EA 14 USD
103807-150 103845-478 76769-436 77082-826 77082-828
Chloromethyl benzoate 98%
Chloromethyl benzoate

Precaución: for Research Use Only

Formula: C₈H₇ClO₂
Boiling Pt: 261.2 °C (760 mm Hg)
Flash Pt: 127.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00040116
CAS Number: 5335-05-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR