Chlorogenic acid 98%

Supplier: Ambeed
Warning

Synonyms: (1S,3R,4R,5R)-3-(((3-(3,4-dihydroxyphenyl)acryloyl)oxy)-1,4,5-trihydroxycyclohexanecarboxylic acid, 3-O-Caffeoylquinic acid, 3-(3,4-Dihydroxycinnamoyl)quinic acid, (1S,3R,4R,5R)-3-[[3-(3,4-Dihydroxyphenyl)-1-oxo-2-propenyl]oxy]-1,4,5-trihydroxycyclohexanecarboxylic acid

A163338-100G A163338-10G A163338-250MG A163338-1G A163338-25G A163338-5G
103805-716EA 17 USD
103805-716 103843-976 77096-214 77096-216 77096-218 77300-652 77300-654 77300-656
Chlorogenic acid 98%
Chlorogenic acid

Precaución: for Research Use Only

Formula: C₁₆H₁₈O₉
MW: 354.31 g/mol
Boiling Pt: 665 °C (760 mm Hg)
Melting Pt: 210 °C
Flash Pt: 245.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00003862
CAS Number: 327-97-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR