(Triphenylphosphine)gold(I) chloride 98%

Supplier: Ambeed
Warning

Synonyms: Chlorotriphenylphosphine gold(I), Chloro(triphenylphosphine)gold(I), Triphenylphosphinechlorogold

A130701-250MG A130701-1G A130701-5G
103801-462EA 77 USD
103801-462 76688-608 77313-918 77313-920
(Triphenylphosphine)gold(I) chloride 98%
(Triphenylphosphine)gold(I) chloride

Precaución: for Research Use Only

Formula: C₁₈H₁₅AuClP
MW: 494.71 g/mol
Boiling Pt: 360 °C (760 mm Hg)
Melting Pt: 248…249 °C
Flash Pt: 181.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009588
CAS Number: 14243-64-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR