CHIR-98014 98%

Supplier: Ambeed
Warning

Synonyms: N2-(2-((4-(2,4-Dichlorophenyl)-5-(1H-imidazol-1-yl)pyrimidin-2-yl)amino)ethyl)-5-nitropyridine-2,6-diamine

A486632-10MG A486632-50MG A486632-5MG A486632-1MG
103791-684EA 78 USD
103791-684 103792-738 103792-740 103799-660 76843-200 76843-202 76843-204 77241-384
CHIR-98014 98%
CHIR-98014

Precaución: for Research Use Only

Formula: C₂₀H₁₇Cl₂N₉O₂
MW: 486.31 g/mol
Storage Temperature: Freezer
MDL Number: MFCD10565922
CAS Number: 252935-94-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR