CGP-53716 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
N-(4-Methyl-3-((4-(pyridin-3-yl)pyrimidin-2-yl)amino)phenyl)benzamide
A128191-1MG
A128191-25MG
A128191-5MG
103791-452EA
52
USD
103791-452
103792-112
103799-326
77271-866
77298-268
77298-270
CGP-53716 98%
CGP-53716
Precaución: for Research Use Only
Formula:
C₂₃H₁₉N₅O MW: 381.42 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00945675 CAS Number: 152459-94-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...