Cephalothin 97%

Supplier: Ambeed
Warning

Synonyms: (6R,7R)-3-(Acetoxymethyl)-8-oxo-7-(2-(thiophen-2-yl)acetamido)-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 7-(2-Thienylacetamido)cephalosporanic Acid, Cephalotin

A437484-10G A437484-100G A437484-500G
103827-624EA 6 USD
103827-624 103838-690 77601-892 77668-958 77668-960
Cephalothin 97%
Cephalothin

Precaución: Keep in a dark place, sealed in dry, room temperature

Formula: C₁₆H₁₆N₂O₆S₂
Boiling Pt: 757.2 °C (760 mm Hg)
Melting Pt: 160…161 °C
Flash Pt: 411.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00242614
CAS Number: 153-61-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR