Zinc(II) (6R,7R)-3-(acetoxymethyl)-7-((R)-5-amino-5-carboxylatopentanamido)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate ≥98%, Ambeed.Inc

Supplier: Ambeed
Danger

Synonyms: Cephalosporin C zinc salt

A164198-100MG A164198-1MG A164198-250MG
103792-236EA 11 USD
103792-236 103799-406 103805-828 103844-088 77611-598 77611-600 77683-354
Zinc(II) (6R,7R)-3-(acetoxymethyl)-7-((R)-5-amino-5-carboxylatopentanamido)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate ≥98%, Ambeed.Inc
Zinc(II) (6R,7R)-3-(acetoxymethyl)-7-((R)-5-amino-5-carboxylatopentanamido)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate

Precaución: For research use only.

Formula: C₁₆H₁₉N₃O₈SZn
Melting Pt: 240…242 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00153937
CAS Number: 59143-60-1
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR