N-Cbz-D-phenylalaninol 97%

Supplier: Ambeed
Warning

Synonyms: N-Benzyloxycarbonyl-D-phenylalaninol, Z-D-Phenylalaninol, Cbz-D-Phenylalaninol

A685189-1G A685189-25G
103840-556EA 44 USD
103840-556 76844-770 76844-772
N-Cbz-D-phenylalaninol 97%
N-Cbz-D-phenylalaninol

Precaución: for Research Use Only

Formula: C₁₇H₁₉NO₃
Boiling Pt: 489 °C (760 mm Hg)
Melting Pt: 92…95 °C
Storage Temperature: Ambient
MDL Number: MFCD00191193
CAS Number: 58917-85-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR