N-Carbobenzoxy-β-alanine 98%

Supplier: Ambeed
Warning

Synonyms: Carbobenzyloxy-beta-alanine

A574726-25G A574726-5G A574726-100G A574726-10G A574726-1G A574726-500G
103839-970EA 8 USD
103839-970 76844-100 76844-102 77280-126 77280-128 77280-130 77280-132
N-Carbobenzoxy-β-alanine 98%
N-Carbobenzoxy-beta-alanine

Precaución: for Research Use Only

Formula: C₁₁H₁₃NO₄
MW: 223.23 g/mol
Boiling Pt: 435.9 °C (760 mm Hg)
Flash Pt: 217.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00037292
CAS Number: 2304-94-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR