Carbenicillin disodium salt ≥770 u/mg

Supplier: Ambeed
Danger

A202423-10G A202423-250MG A202423-5G
103817-386EA 10 USD
103817-386 103848-026 77685-513 77685-514 77685-515
Carbenicillin disodium salt ≥770 u/mg
Carbenicillin disodium salt

Precaución: for Research Use Only

Formula: C₁₇H₁₆N₂O₆S·2Na
Storage Temperature: Refrigerator
MDL Number: MFCD00077683
CAS Number: 4800-94-6
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR