Chloroformamidine hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: Carbamimidic chloride hydrochloride

A189277-100G A189277-10G A189277-250MG A189277-500G A189277-1G A189277-5G
103816-174EA 6 USD
103816-174 103846-768 76690-816 76690-818 76690-820 76690-822 77084-534 77084-536
Chloroformamidine hydrochloride 97%
Chloroformamidine hydrochloride

Precaución: for Research Use Only

Formula: CH₄Cl₂N₂
MW: 114.96 g/mol
Melting Pt: 176…178 °C
Storage Temperature: Ambient
MDL Number: MFCD00035527
CAS Number: 29671-92-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR