Candesartan 95%

Supplier: Ambeed
Danger

Synonyms: 1-((2'-(2H-Tetrazol-5-yl)-[1,1'-biphenyl]-4-yl)methyl)-2-ethoxy-1H-benzo[d]imidazole-7-carboxylic acid, 1-2-1H-Tetrazol-5-yl-1-1-biphenyl-4-yl-methyl-2-ethoxy-1H-benzo-d-imidazole-7-carboxylic-acid, 1-((2'-(1H-Tetrazol-5-yl)[1,1'-biphenyl]-4-yl)methyl)-2-ethoxy-1H-benzo[d]imidazole-7-carboxylic acid

A148921-1G A148921-5G A148921-10MG
103803-982EA 23 USD
103803-982 103842-102 76997-256 77317-570 77317-572 77671-092
Candesartan 95%
Candesartan

Precaución: for Research Use Only

Formula: C₂₄H₂₀N₆O₃
Boiling Pt: 754.8 °C (760 mm Hg)
Melting Pt: 183…185 °C
Storage Temperature: Freezer
MDL Number: MFCD00864463
CAS Number: 139481-59-7
UN: 1325
ADR: 4.1,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR