Adenosine-3',5'-cyclic monophosphate (cAMP) 99%
Supplier: Ambeed
|
Danger
|
Synonyms:
3',5'-cAMP, 3′,5′-Cyclic AMP, Adenosine 3′,5′-cyclophosphate, (4aR,6R,7R,7aS)-6-(6-Amino-9H-purin-9-yl)-2,7-dihydroxytetrahydro-4H-furo[3,2-d][1,3,2]dioxaphosphinine 2-oxide
A353547-100MG
A353547-5G
A353547-250MG
103826-098EA
23
USD
103826-098
103837-100
76694-116
76840-214
77626-684
Adenosine-3',5'-cyclic monophosphate (cAMP) 99%
Adenosine-3',5'-cyclic monophosphate (cAMP)
Precaución: for Research Use Only
Formula:
C₁₀H₁₂N₅O₆P MW: 329.21 g/mol Boiling Pt: 701.5 °C (760 mm Hg) Melting Pt: 260 °C Storage Temperature: Freezer |
MDL Number:
MFCD00005845 CAS Number: 60-92-4 UN: 3261 ADR: 8,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...