Caffeic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3,4-Dihydroxycinnamic acid, 3-(3,4-Dihydroxyphenyl)acrylic acid

A134210-100G A134210-10G A134210-500G A134210-5G A134210-25G A134210-1000G
76688-746EA 51.99 USD
76688-746 76688-748 76688-750 77298-696 77314-708 77624-126
Caffeic acid 98%
Caffeic acid

Precaución: for Research Use Only

Formula: C₉H₈O₄
MW: 180.16 g/mol
Boiling Pt: 416.8 °C (760 mm Hg)
Melting Pt: 211…213 °C
Flash Pt: 220 °C
Storage Temperature: Ambient
MDL Number: MFCD00004392
CAS Number: 331-39-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR