Butyl-p-toluenesulfonate 96%
Supplier: Ambeed
|
Warning
|
Synonyms:
Butyl 4-methylbenzenesulphonate, p-Toluenesulphonic acid Butyl Ester
A167649-10G
A167649-25G
A167649-5G
103844-464EA
7
USD
103844-464
76720-376
77081-488
77081-490
Butyl-p-toluenesulfonate 96%
Butyl-p-toluenesulfonate
Precaución: for Research Use Only
Formula:
C₁₁H₁₆O₃S MW: 228.31 g/mol Boiling Pt: 335.4 °C (760 mm Hg) Flash Pt: 156.6 °C Storage Temperature: Ambient |
MDL Number:
MFCD00027203 CAS Number: 778-28-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...