Butyl 2-cyanoacrylate 95% stabilized

Supplier: Ambeed
Warning

Synonyms: Enbucrilate

Stabilization: Stabilized with TBC or HQ

A224898-250MG A224898-5G
103819-142EA 26 USD
103819-142 103850-034 76800-944 77088-698
Butyl 2-cyanoacrylate 95% stabilized
Butyl 2-cyanoacrylate

Precaución: for Research Use Only

Formula: C₈H₁₁NO₂
MW: 153.17 g/mol
Boiling Pt: 232.8 °C (760 mm Hg)
Flash Pt: 100.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00134196
CAS Number: 6606-65-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR