3-Butynoic acid 98%

Supplier: Ambeed
Danger

Synonyms: But-3-ynoic acid

A420783-250MG A420783-10G
103827-394EA 25 USD
103827-394 103838-442 77616-826 77748-851
3-Butynoic acid 98%
3-Butynoic acid

Precaución: Keep in a dark place, inert atmosphere, store in freezer, under -20 °C

Formula: C₄H₄O₂
MW: 84.07 g/mol
Boiling Pt: 194.9 °C (760 mm Hg)
Melting Pt: 70…80 °C
Flash Pt: 82.6 °C
Storage Temperature: Freezer
MDL Number: MFCD02258494
CAS Number: 2345-51-9
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR