3-Butyn-1-amine hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: 3-Butynylamine hydrochloride, 3-Butyn-1-ylammonium chloride, But-3-yn-1-ylammonium chloride, But-3-yn-1-amine hydrochloride

A196114-1G A196114-25G A196114-5G A196114-10G A196114-250MG A196114-100G
103816-814EA 23 USD
103816-814 103847-428 77085-378 77085-380 77085-382 77152-012 77152-014 77683-535
3-Butyn-1-amine hydrochloride 95%
3-Butyn-1-amine hydrochloride

Precaución: For research use only.

Formula: C₄H₇N·HCl
MW: 105.57 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06658391
CAS Number: 88211-50-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR