But-1-en-2-ylbenzene 95% stabilized

Supplier: Ambeed
Warning


Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

A200302-250MG A200302-1G
103817-188EA 220 USD
103817-188 103847-816 77274-790 77302-674
But-1-en-2-ylbenzene 95% stabilized
But-1-en-2-ylbenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₂
MW: 132.2 g/mol
Boiling Pt: 184.8 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD00060866
CAS Number: 2039-93-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR