(Methoxycarbonylsulfamoyl)triethylammonium hydroxide inner salt 95%

Supplier: Ambeed
Warning

A699984-1G A699984-25G A699984-5G A699984-10G A699984-100G A699984-500G A699984-1000G
103840-630EA 95 USD
103840-630 76697-312 76844-852 76844-854 76844-856 77552-122 77569-332 77602-776 77627-430
(Methoxycarbonylsulfamoyl)triethylammonium hydroxide inner salt 95%
(Methoxycarbonylsulfamoyl)triethylammonium hydroxide inner salt

Precaución: For research use only.

Formula: C₈H₁₈N₂O₄S
Melting Pt: 76…79 °C
Storage Temperature: Freezer
MDL Number: MFCD00077815
CAS Number: 29684-56-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR