Buflomedil hydrochloride 98%

Supplier: Ambeed
Warning

A113192-100MG A113192-25MG A113192-1G A113192-10G A113192-25G A113192-5G A113192-100G
103798-182EA 18 USD
103798-182 77148-812 77148-814 77297-228 77685-002 77685-003 77685-004 77900-512
Buflomedil hydrochloride 98%
Buflomedil hydrochloride

Precaución: for Research Use Only

Formula: C₁₇H₂₅NO₄
Boiling Pt: 454.5 °C (760 mm Hg)
Flash Pt: 228.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00078965
CAS Number: 35543-24-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR