Bromotrispyrrolidinophosphonium hexafluorophosphate 98%

Supplier: Ambeed
Danger

Synonyms: Bromotri(1-pyrrolidinyl)phosphonium hexafluorophosphate, PyBrOP , Bromotripyrrolidinylphosphonium hexafluorophosphate, Bromotri(pyrrolidin-1-yl)phosphonium hexafluorophosphate(V)

A173610-10G A173610-25G A173610-5G A173610-1G A173610-100G
103806-840EA 8 USD
103806-840 103845-140 76998-306 77082-390 77082-392 77301-286 77881-332
Bromotrispyrrolidinophosphonium hexafluorophosphate 98%
Bromo-tris-pyrrolidinophosphonium hexafluorophosphate

Precaución: for Research Use Only

Formula: C₁₂H₂₄BrF₆N₃P₂
MW: 466.2 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00077412
CAS Number: 132705-51-2
UN: 3077
ADR: 9 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR