5-Bromo-2'-deoxyuridine (broxuridine) 98%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-1-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione, 5-Bromo-2'-deoxyuridine, 5-BrdU

A386942-100MG A386942-250MG A386942-1G A386942-5G A386942-10G A386942-100G
103826-810EA 18 USD
103826-810 103837-852 76817-732 76817-734 76841-210 77306-978 77601-704 77672-916
5-Bromo-2'-deoxyuridine (broxuridine) 98%
5-Bromo-2'-deoxyuridine (broxuridine)

Precaución: for Research Use Only

Formula: C₉H₁₁BrN₂O₅
MW: 307.10 g/mol
Melting Pt: 191…194 °C
Storage Temperature: Freezer
MDL Number: MFCD00006529
CAS Number: 59-14-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR