Bromhexine hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-3,5-dibromo-N-cyclohexyl-N-methylbenzylamine Hydrochloride, 2,4-Dibromo-6-((cyclohexyl(methyl)amino)methyl)aniline hydrochloride, N-(2-Amino-3,5-dibromobenzyl)-N-methylcyclohexylamine Hydrochloride

A128682-1000G A128682-100G A128682-500G A128682-5G
77109-066EA 253.38 USD
77109-066 77109-068 77109-070 77313-518
Bromhexine hydrochloride 97%
Bromhexine hydrochloride

Precaución: for Research Use Only

Formula: C₁₄H₂₀Br₂N₂
Melting Pt: 240…245 °C
Flash Pt: 220.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00056626
CAS Number: 611-75-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR