N-(tert-Butoxycarbonyl)-6-aminohexanoic acid 98%

Supplier: Ambeed
Warning

Synonyms: 6-[(tert-Butoxycarbonyl)amino]hexanoic Acid, 6-(Boc-amino)hexanoic Acid, Boc-6-Ahx-OH, N-Boc-6-aminohexanoic Acid, Boc-ε-Acp-OH

A704326-100G A704326-10G A704326-25G A704326-5G A704326-500G
103840-660EA 5 USD
103840-660 76820-048 76820-050 77308-166 77308-168 77673-924
N-(tert-Butoxycarbonyl)-6-aminohexanoic acid 98%
N-(tert-Butoxycarbonyl)-6-aminohexanoic acid

Precaución: for Research Use Only

Formula: C₁₁H₂₁NO₄
MW: 231.29 g/mol
Melting Pt: 35…40 °C
Flash Pt: 183.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00037798
CAS Number: 6404-29-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR