(R)-Methyl-2-((tert-butoxycarbonyl)amino)-3-iodopropanoate 98%

Supplier: Ambeed
Warning

Synonyms: Boc-β-iodo-Ala-OMe, BOC-3-iodo-L-alanine methyl ester, N-Boc-3-iodo-L-alanine Methyl Ester, Boc-β-Iodo-Ala-OMe, Boc-Ala(3-I)-OMe

A187373-10G A187373-250MG A187373-1G A187373-25G A187373-100G A187373-500G
103816-006EA 6 USD
103816-006 103846-596 76872-962 76872-964 77084-284 77084-286 77550-198 77611-820
(R)-Methyl-2-((tert-butoxycarbonyl)amino)-3-iodopropanoate 98%
(R)-Methyl-2-((tert-butoxycarbonyl)amino)-3-iodopropanoate

Precaución: for Research Use Only

Formula: C₉H₁₆INO₄
MW: 329.13 g/mol
Boiling Pt: 356.5 °C (760 mm Hg)
Melting Pt: 50…52 °C
Flash Pt: 169.4 °C
Storage Temperature: Freezer
MDL Number: MFCD00216579
CAS Number: 93267-04-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR