3-(tert-Butoxycarbonylamino)-1-propanol 97%

Supplier: Ambeed
Warning

Synonyms: 3-(Boc-amino)-1-propanol, N-(3-Hydroxypropyl)carbamic Acid tert-Butyl Ester, Boc-β-Ala-ol, 3-(BOC-amino)-1-propanol, tert-Butyl N-(3-Hydroxypropyl)carbamate

A199657-25G A199657-5G A199657-100G A199657-10G A199657-500G A199657-1G A199657-1000G
103817-122EA 6 USD
103817-122 103847-748 77085-794 77085-796 77274-770 77274-772 77274-774 77302-624 77615-694
3-(tert-Butoxycarbonylamino)-1-propanol 97%
3-(tert-Butoxycarbonylamino)-1-propanol

Precaución: for Research Use Only

Formula: C₈H₁₇NO₃
Boiling Pt: 293 °C (760 mm Hg)
Flash Pt: 109 °C
Storage Temperature: Ambient
MDL Number: MFCD00191883
CAS Number: 58885-58-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR