Boc-b-Alanine 97%

Supplier: Ambeed
Warning

Synonyms: N-Boc-beta-alanine, Boc-β-Ala-OH, BOC-beta-alanine

A101626-100G A101626-10G A101626-500G A101626-25G A101626-1000G A101626-5G
103807-808EA 91 USD
103807-808 103821-746 76970-178 76970-180 76970-182 76994-522 77297-174 77297-176
Boc-b-Alanine 97%
Boc-b-Alanine

Precaución: for Research Use Only

Formula: C₈H₁₅NO₄
MW: 189.2 g/mol
Boiling Pt: 333.9 °C (760 mm Hg)
Melting Pt: 76…78 °C
Flash Pt: 155.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00037291
CAS Number: 3303-84-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR