Boc-L-valine hydroxysuccinimide ester 97%

Supplier: Ambeed
Warning

Synonyms: N-Boc-L-valine N-succinimidyl ester, Boc-Val-OSu

A340967-10G A340967-25G A340967-5G
103836-800EA 10 USD
103836-800 76773-124 77306-528 77306-530
Boc-L-valine hydroxysuccinimide ester 97%
Boc-L-valine hydroxysuccinimide ester

Precaución: for Research Use Only

Formula: C₁₄H₂₂N₂O₆
MW: 314.338 g/mol
Melting Pt: 126…128 °C
Storage Temperature: Freezer
MDL Number: MFCD00037906
CAS Number: 3392-12-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR