N-Boc-L-valine 97%

Supplier: Ambeed
Warning

Synonyms: Boc-Val-OH, BOC-L-Valine

A175080-25G A175080-100G A175080-500G A175080-1000G
103845-300EA 6 USD
103845-300 77082-620 77273-844 77273-846 77273-848 77615-458
N-Boc-L-valine 97%
N-Boc-L-valine

Precaución: for Research Use Only

Formula: C₁₀H₁₉NO₄
Boiling Pt: 341.8 °C (760 mm Hg)
Melting Pt: 77…80 °C
Flash Pt: 160.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00065605
CAS Number: 13734-41-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR