tert-Butyl (1S)-1-(hydroxymethyl)-2-methylpropylcarbamate 98%

Supplier: Ambeed
Warning

Synonyms: N-BOC-L-Valinol, Boc-Valinol, (S)-tert-butyl (1-hydroxy-3-methylbutan-2-yl)carbamate

A310965-10G A310965-1G A310965-25G
103814-644EA 9 USD
103814-644 103835-830 76772-782 76838-608 76838-610
tert-Butyl (1S)-1-(hydroxymethyl)-2-methylpropylcarbamate 98%
tert-Butyl (1S)-1-(hydroxymethyl)-2-methylpropylcarbamate

Precaución: for Research Use Only

Formula: C₁₀H₂₁NO₃
MW: 203.28 g/mol
Boiling Pt: 208 °C (760 mm Hg)
Flash Pt: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD00082635
CAS Number: 79069-14-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR