Boc-Tyr-OtBu 98%

Supplier: Ambeed
Warning

A175722-5G A175722-100G A175722-10G A175722-25G A175722-500G
103807-082EA 6 USD
103807-082 103845-394 77082-738 77151-476 77151-478 77301-402 77685-394
Boc-Tyr-OtBu 98%
Boc-Tyr-OtBu

Precaución: for Research Use Only

Formula: C₁₈H₂₇NO₅
MW: 337.41 g/mol
Boiling Pt: 476.0±40.0 °C (760 mm Hg)
Flash Pt: 241.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD20264826
CAS Number: 18938-60-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR