N-(tert-Butoxycarbonyl)-O-benzyl-L-tyrosine 98%

Supplier: Ambeed
Warning

Synonyms: N-Boc-O-benzyl-L-tyrosine, BOC-O-Benzyl-L-tyrosine, Boc-Tyr(Bzl)-OH

A105900-100G A105900-10G A105900-25G A105900-1G A105900-5G
103796-964EA 9 USD
103796-964 103822-710 77270-530 77270-532 77270-534 77309-572 77309-574
N-(tert-Butoxycarbonyl)-O-benzyl-L-tyrosine 98%
N-(tert-Butoxycarbonyl)-O-benzyl-L-tyrosine

Precaución: for Research Use Only

Formula: C₂₁H₂₅NO₅
MW: 371.43 g/mol
Boiling Pt: 552.4 °C (760 mm Hg)
Melting Pt: 110…112 °C
Flash Pt: 287.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00065597
CAS Number: 2130-96-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR