N-ɑ-Boc-L-tryptophanol 98%

Supplier: Ambeed
Warning

Synonyms: (S)-2-(tert-Butoxycarbonyl)amino-3-(3-indolyl)propanol, N-ɑ-(tert-Butoxycarbonyl)-L-tryptophanol, (S)-2-(Boc-amino)-3-(3-indolyl)propanol, Boc-Tryptophanol, N-Boc-L-tryptophanol

A221047-100G A221047-10G A221047-1G A221047-25G A221047-5G
103818-886EA 7 USD
103818-886 103849-766 76873-956 76873-958 77088-356 77088-358 77088-360
N-ɑ-Boc-L-tryptophanol 98%
N-alpha-Boc-L-tryptophanol

Precaución: for Research Use Only

Formula: C₁₆H₂₂N₂O₃
MW: 290.36 g/mol
Boiling Pt: 518.1 °C (760 mm Hg)
Melting Pt: 118…122 °C
Storage Temperature: Ambient
MDL Number: MFCD00235953
CAS Number: 82689-19-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR